Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdyl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123386.1 | NP_001116858.1 | 4 - 5 | 1581 | 136 | |
NM_009881.3 | NP_034011.1 | 7 - 8 | 1531 | 136 | ||
GenBank mRNA | AF081260.1 | - | 7 - 8 | 1513 | 136 | |
AF081261.1 | - | 7 - 8 | 1513 | 136 | ||
AK028162.1 | - | 4 - 5 | 1277 | 136 | ||
AK156509.1 | - | 4 - 5 | 1582 | 136 | ||
AK162180.1 | - | 2 - 3 | 261 | 136 | ||
BC055103.1 | - | 7 - 8 | 1480 | 6741468 | 136 | |
BC062123.1 | - | 4 - 5 | 1670 | 6826210 | 136 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |