Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zmat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175446.3 | NP_780655.2 | 2 - 3 | 403 | 102 | |
GenBank mRNA | AK133258.1 | - | 355 | 102 | ||
AK220419.1 | - | 1 - 2 | 44 | 102 | ||
AW044958.1 | - | 2 - 3 | 405 | 102 | ||
BC067022.1 | - | 2 - 3 | 379 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |