Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trak1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175114.3 | NP_780323.2 | 2 - 3 | 434 | 95 | |
GenBank mRNA | AK029123.1 | - | 3 - 4 | 832 | 95 | |
AK041436.1 | - | 3 - 4 | 802 | 95 | ||
AK052264.1 | - | 3 - 4 | 821 | 95 | ||
AK122426.1 | - | 3 - 4 | 524 | 95 | ||
BC046285.1 | - | 2 - 3 | 424 | 95 | ||
BC058971.1 | - | 2 - 3 | 376 | 6832507 | 95 | |
BC116262.1 | - | 2 - 3 | 401 | 95 | ||
BC116263.1 | - | 2 - 3 | 401 | 95 | ||
BC129961.1 | - | 2 - 3 | 401 | 95 | ||
BC129962.1 | - | 2 - 3 | 401 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |