Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172520.2 | NP_766108.1 | 12 - 13 | 2046 | 74 | |
GenBank mRNA | AK031443.1 | - | 12 - 13 | 2047 | 74 | |
AK053075.1 | - | 12 - 13 | 2047 | 74 | ||
BC060376.1 | - | 10 - 11 | 1776 | 5340007 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |