Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chodl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139134.3 | NP_624360.2 | 3 - 4 | 964 | 70 | |
GenBank mRNA | AK014255.1 | - | 3 - 4 | 675 | 70 | |
AK031063.1 | - | 3 - 4 | 965 | 70 | ||
AK076523.1 | - | 3 - 4 | 964 | 70 | ||
AK140910.1 | - | 3 - 4 | 965 | 70 | ||
BC117071.1 | - | 3 - 4 | 635 | 40126390 | 70 | |
BC117073.1 | - | 3 - 4 | 635 | 40126392 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |