Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Poglut1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300827.1 | NP_001287756.1 | 8 - 9 | 1082 | 73 | |
NM_172380.4 | NP_759012.1 | 8 - 9 | 879 | 73 | ||
GenBank mRNA | AK031608.1 | - | 8 - 9 | 818 | 73 | |
AK035948.1 | - | 8 - 9 | 827 | 73 | ||
AK036224.1 | - | 8 - 9 | 809 | 73 | ||
AK041321.1 | - | 8 - 9 | 817 | 73 | ||
AK045278.1 | - | 8 - 9 | 1027 | 73 | ||
AK079886.1 | - | 8 - 9 | 849 | 73 | ||
BC026809.1 | - | 8 - 9 | 830 | 4205469 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |