Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tdrd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290475.1 | NP_001277404.1 | 15 - 16 | 3028 | 70 | |
NM_146142.2 | NP_666254.1 | 15 - 16 | 3026 | 70 | ||
GenBank mRNA | AK150077.1 | - | 15 - 16 | 2926 | 70 | |
BC025099.1 | - | 3 - 4 | 682 | 70 | ||
BC029689.1 | - | 14 - 15 | 2900 | 5367741 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |