Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Suv39h1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290716.1 | NP_001277645.1 | 3 - 4 | 1003 | 104 | |
NM_011514.2 | NP_035644.1 | 3 - 4 | 1022 | 104 | ||
GenBank mRNA | AF019969.1 | - | 3 - 4 | 848 | 104 | |
AF193861.1 | - | 3 - 4 | 844 | 104 | ||
AF193862.1 | - | 3 - 4 | 947 | 104 | ||
AK088405.1 | - | 3 - 4 | 839 | 104 | ||
AK132238.1 | - | 3 - 4 | 884 | 104 | ||
AK169389.1 | - | 3 - 4 | 830 | 104 | ||
AK169592.1 | - | 3 - 4 | 992 | 104 | ||
BC023860.1 | - | 3 - 4 | 899 | 5352230 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |