Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufv3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083891.1 | NP_001077360.1 | 2 - 3 | 274 | 68 | |
GenBank mRNA | AK077433.1 | - | 2 - 3 | 215 | 68 | |
BU555486.1 | - | 2 - 3 | 277 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Transporter 3' Most Mus musculus Probe spans exons |