Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrto4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290810.1 | NP_001277739.1 | 7 - 8 | 846 | 124 | |
GenBank mRNA | AK011387.1 | - | 7 - 8 | 568 | 124 | |
BC096556.1 | - | 7 - 8 | 589 | 124 | ||
BC138221.1 | - | 7 - 8 | 645 | 8861240 | 124 | |
BC145205.1 | - | 7 - 8 | 645 | 9053756 | 124 | |
BC145209.1 | - | 7 - 8 | 667 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |