Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iba57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173785.6 | NP_776146.1 | 1 - 2 | 393 | 78 | |
GenBank mRNA | AK038625.1 | - | 1 - 2 | 388 | 78 | |
AK133203.1 | - | 1 - 2 | 391 | 78 | ||
AK172525.1 | - | 1 - 2 | 354 | 78 | ||
BC094909.1 | - | 1 - 2 | 393 | 6432747 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |