Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fktn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139309.4 | NP_647470.1 | 2 - 3 | 277 | 102 | |
GenBank mRNA | AB077383.1 | - | 3 - 4 | 461 | 102 | |
AJ511807.1 | - | 3 - 4 | 297 | 102 | ||
AK052357.1 | - | 4 - 5 | 435 | 102 | ||
AK085931.1 | - | 2 - 3 | 278 | 102 | ||
AK142744.1 | - | 437 | 102 | |||
AK144743.1 | - | 3 - 4 | 306 | 102 | ||
BC017538.1 | - | 3 - 4 | 293 | 4506804 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |