Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK033805.1 | - | 696 | 74 | ||
BC139325.1 | - | 1 - 2 | 1211 | 8862347 | 74 | |
BC139327.1 | - | 1 - 2 | 1211 | 8862349 | 74 | |
M64226.1 | - | 1 - 2 | 1127 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |