Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hs3st3a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178870.5 | NP_849201.1 | 2 | 3626 | 141 | |
GenBank mRNA | AK032282.1 | - | 2 | 2594 | 141 | |
AK051284.1 | - | 2 | 3619 | 141 | ||
BC094320.1 | - | 2 | 3680 | 6400013 | 141 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |