Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1400018657.mm.1 |
NM_172527.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400018657.mm.1 |
NM_172527.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400018657.mm.1 |
TC1400002395.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400018660.mm.1 |
NM_172527.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400018660.mm.1 |
NM_172527.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400018660.mm.1 |
TC1400002395.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400018659.mm.1 |
NM_172527.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400018659.mm.1 |
NM_172527.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400018659.mm.1 |
TC1400002395.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400018656.mm.1 |
NM_172527.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400018656.mm.1 |
NM_172527.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400018656.mm.1 |
TC1400002395.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400018658.mm.1 |
NM_172527.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400018658.mm.1 |
NM_172527.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400018658.mm.1 |
TC1400002395.mm.1 |
MTA TC |