Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jph4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177049.5 | NP_796023.2 | 2 - 3 | 1493 | 106 | |
GenBank mRNA | AB099712.1 | - | 2 - 3 | 836 | 106 | |
AK031092.1 | - | 1 - 2 | 540 | 106 | ||
AK033956.1 | - | 2 - 3 | 1126 | 106 | ||
AK034118.1 | - | 2 - 3 | 1127 | 106 | ||
AK036161.1 | - | 2 - 3 | 1139 | 106 | ||
AK048607.1 | - | 1 - 2 | 594 | 106 | ||
AK148135.1 | - | 1 - 2 | 657 | 106 | ||
AY341261.1 | - | 2 - 3 | 1493 | 106 | ||
AY350744.1 | - | 1 - 2 | 480 | 106 | ||
BC052049.1 | - | 1 - 2 | 538 | 5689347 | 106 | |
BC052376.1 | - | 2 - 3 | 590 | 5696231 | 106 | |
BC052722.1 | - | 2 - 3 | 564 | 6810400 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |