Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Senp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163571.1 | NP_001157043.1 | 9 - 10 | 1656 | 129 | |
NM_030702.4 | NP_109627.3 | 9 - 10 | 1868 | 129 | ||
GenBank mRNA | AF194031.1 | - | 8 - 9 | 1549 | 129 | |
AK045071.1 | - | 9 - 10 | 1745 | 129 | ||
AK088608.1 | - | 9 - 10 | 1657 | 129 | ||
AY008764.1 | - | 9 - 10 | 1600 | 129 | ||
BC037014.1 | - | 9 - 10 | 1634 | 5344274 | 129 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |