Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008163.3 | NP_032189.1 | 3 - 4 | 506 | 107 | |
GenBank mRNA | AK077750.1 | - | 2 - 3 | 325 | 107 | |
AK079087.1 | - | 2 - 3 | 108 | |||
AK148148.1 | - | 3 - 4 | 471 | 107 | ||
AK153557.1 | - | 2 - 3 | 439 | 107 | ||
AK155800.1 | - | 3 - 4 | 461 | 107 | ||
AK166142.1 | - | 3 - 4 | 460 | 107 | ||
AK170845.1 | - | 2 - 3 | 297 | 107 | ||
AK172002.1 | - | 3 - 4 | 108 | |||
BC052377.1 | - | 3 - 4 | 506 | 5696248 | 107 | |
BC085254.1 | - | 3 - 4 | 342 | 5343274 | 107 | |
U07617.1 | - | 2 - 3 | 255 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Probe spans exons |