Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Larp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024526.2 | NP_001019697.2 | 12 - 13 | 1559 | 62 | |
NM_001080948.2 | NP_001074417.1 | 12 - 13 | 1572 | 62 | ||
NM_001284521.1 | NP_001271450.1 | 12 - 13 | 1546 | 62 | ||
NM_001284522.1 | NP_001271451.1 | 12 - 13 | 1569 | 62 | ||
NM_001284523.1 | NP_001271452.1 | 12 - 13 | 1562 | 62 | ||
GenBank mRNA | AK049756.1 | - | 12 - 13 | 1486 | 62 | |
AK139636.1 | - | 12 - 13 | 1567 | 62 | ||
AK139909.1 | - | 12 - 13 | 1410 | 62 | ||
AK165392.1 | - | 12 - 13 | 1506 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |