Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rmrp | |||||||
---|---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_001460.1 | LIT3323 RMRP |
1 | 148 | 111 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | suppression inducing transmembrane adaptor 1 | Sit1 | Sit | -9.1kb | Sense | Chr. 4 - 43482083 - 43483709 on NCBI Build GRCm38 |
Downstream | coiled-coil domain containing 107 | Ccdc107 | 281.0 | Anti-sense | Chr. 4 - 43493340 - 43495921 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |