Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snhg12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK009175.1 | - | 3 - 4 | 364 | 97 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
small nucleolar RNA, H/ACA box 61 |
Snora61 | MBI-164 | Sense | Chr. 4 - 132310307 - 132310368 on NCBI Build GRCm38 | |
Downstream | tRNA selenocysteine 1 associated protein 1 | Trnau1ap |
1110007F05Rik, AA387999, Secp43, Trspap1, WI1-2643G14.2 |
742.0 | Anti-sense | Chr. 4 - 132311763 - 132329545 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons |