Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C330019O22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK021212.1 | - | 1 | 540 | 96 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | tRNA methyltransferase 10C | Trmt10c |
1300018J16Rik, D16Ertd454e, Rg9mtd1, Rnmtd1 |
-77.5kb | Sense | Chr. 16 - 56033720 - 56037774 on NCBI Build GRCm38 |
Downstream | interphotoreceptor matrix proteoglycan 2 | Impg2 |
IPM200, PG10.2, Rsbp, Spacrcan |
87.6kb | Anti-sense | Chr. 16 - 56203842 - 56273753 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |