Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rybp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019743.3 | NP_062717.2 | 3 - 4 | 413 | 80 | |
GenBank mRNA | AF101779.1 | - | 3 - 4 | 470 | 80 | |
AK033024.1 | - | 3 - 4 | 452 | 80 | ||
BC053016.1 | - | 3 - 4 | 413 | 6406584 | 80 | |
BC080287.1 | - | 3 - 4 | 496 | 6410301 | 80 | |
BC138324.1 | - | 3 - 4 | 395 | 8861344 | 80 | |
BC138325.1 | - | 3 - 4 | 395 | 8861345 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Probe spans exons |