Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pard6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047435.2 | NP_001040900.1 | 2 - 3 | 382 | 101 | |
GenBank mRNA | AK003030.1 | - | 2 - 3 | 351 | 101 | |
AK158848.1 | - | 2 - 3 | 380 | 101 | ||
BC049593.1 | - | 2 - 3 | 359 | 6771847 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |