Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103171.1 | NP_001096641.1 | 51 - 52 | 9263 | 106 | |
NM_010862.2 | NP_034992.2 | 52 - 53 | 9317 | 106 | ||
NM_182698.2 | NP_874357.2 | 50 - 51 | 5501 | 106 | ||
GenBank mRNA | AF144095.1 | - | 52 - 53 | 9317 | 106 | |
AK030528.1 | - | 2 - 3 | 297 | 106 | ||
AY331132.1 | - | 51 - 52 | 9263 | 106 | ||
AY331133.1 | - | 50 - 51 | 5501 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |