Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400008773.mm.1 |
NM_001166027.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008773.mm.1 |
NM_001166027.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008773.mm.1 |
NM_177669.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008773.mm.1 |
NM_177669.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008773.mm.1 |
TC0400001207.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400008775.mm.1 |
NM_001166027.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008775.mm.1 |
NM_001166027.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008775.mm.1 |
TC0400001207.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400008770.mm.1 |
NM_001166027.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008770.mm.1 |
NM_001166027.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008770.mm.1 |
TC0400001207.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400008772.mm.1 |
NM_001166027.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008772.mm.1 |
NM_001166027.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008772.mm.1 |
NM_177669.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008772.mm.1 |
NM_177669.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008772.mm.1 |
TC0400001207.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400008771.mm.1 |
NM_001166027.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008771.mm.1 |
NM_001166027.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400008771.mm.1 |
TC0400001207.mm.1 |
MTA TC |