Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cxcl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008176.3 | NP_032202.1 | 2 - 3 | 258 | 58 | |
GenBank mRNA | BC037997.1 | - | 2 - 3 | 225 | 58 | |
BC132502.1 | - | 2 - 3 | 217 | 40130779 | 58 | |
BC132504.1 | - | 2 - 3 | 217 | 40130781 | 58 | |
BQ031102.1 | - | 2 - 3 | 58 | |||
BY335927.1 | - | 2 - 3 | 265 | 58 | ||
J04596.1 | - | 2 - 3 | 193 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |