Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcm7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008568.2 | NP_032594.1 | 11 - 12 | 1701 | 111 | |
GenBank mRNA | AK076059.1 | - | 7 - 8 | 1151 | 111 | |
AK077766.1 | - | 8 - 9 | 1260 | 111 | ||
AK132748.1 | - | 2265 | 111 | |||
AK133091.1 | - | 11 - 12 | 1697 | 111 | ||
AK144980.1 | - | 11 - 12 | 1713 | 111 | ||
AK149913.1 | - | 11 - 12 | 1695 | 111 | ||
AK150059.1 | - | 11 - 12 | 1698 | 111 | ||
AK150066.1 | - | 11 - 12 | 1695 | 111 | ||
AK154052.1 | - | 11 - 12 | 1698 | 111 | ||
BC023232.1 | - | 11 - 12 | 1677 | 111 | ||
BC065164.1 | - | 11 - 12 | 1685 | 6829737 | 111 | |
BC066024.1 | - | 11 - 12 | 1640 | 6829337 | 111 | |
D26091.1 | - | 11 - 12 | 1701 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |