Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sost | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024449.6 | NP_077769.4 | 1 - 2 | 263 | 84 | |
GenBank mRNA | AK017295.1 | - | 1 - 2 | 264 | 84 | |
AK041382.1 | - | 1 - 2 | 264 | 84 | ||
BC137832.1 | - | 1 - 2 | 253 | 8860851 | 84 | |
BC137833.1 | - | 1 - 2 | 253 | 8860852 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |