Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Noxo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027988.4 | NP_082264.2 | 4 - 5 | 1237 | 111 | |
GenBank mRNA | AF399754.1 | - | 3 - 4 | 223 | 111 | |
AF539797.1 | - | 3 - 4 | 223 | 111 | ||
AK009605.1 | - | 3 - 4 | 332 | 111 | ||
AK088226.1 | - | 4 - 5 | 1196 | 111 | ||
AK157993.1 | - | 4 - 5 | 1238 | 111 | ||
BC019525.1 | - | 3 - 4 | 249 | 4220018 | 111 | |
BC119527.1 | - | 3 - 4 | 325 | 111 | ||
BC127235.1 | - | 3 - 4 | 325 | 111 | ||
BC127236.1 | - | 3 - 4 | 325 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |