Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Neu4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173772.3 | NP_776133.1 | 3 - 4 | 611 | 81 | |
GenBank mRNA | AB510404.1 | - | 4 - 5 | 592 | 81 | |
AK034236.1 | - | 3 - 4 | 582 | 81 | ||
AY258421.1 | - | 3 - 4 | 524 | 81 | ||
BC070438.1 | - | 3 - 4 | 611 | 6842155 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |