Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024175.3 | NP_077137.1 | 1 | 44 | 83 | |
GenBank mRNA | AK166994.1 | - | 1 | 36 | 83 | |
BY043509.1 | - | 1 | 44 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Amplicon spans exons and probe does not span exons |