Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200005433.mm.1 |
NM_001252547.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005433.mm.1 |
NM_001252547.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005433.mm.1 |
NM_013781.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005433.mm.1 |
NM_013781.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005433.mm.1 |
NR_045535.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005433.mm.1 |
NR_045535.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005433.mm.1 |
TC0200000591.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200005431.mm.1 |
NR_045535.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005431.mm.1 |
NR_045535.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200005431.mm.1 |
TC0200000591.mm.1 |
MTA TC |