Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_073203.1 | 1 - 2 | 146 | 107 | ||
GenBank mRNA | BC089482.1 | - | 145 | 107 | ||
BC099544.1 | - | 1 - 2 | 140 | 107 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | sprouty homolog 2 (Drosophila) | Spry2 | sprouty2 | -209.4kb | Anti-sense | Chr. 14 - 105891947 - 105896819 on NCBI Build GRCm38 |
Downstream | RIKEN cDNA 1700128A07 gene | 1700128A07Rik | 281.3kb | Anti-sense | Chr. 14 - 106417187 - 106486520 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |