Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BMP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110277.1 | NP_001103747.1 | 2 - 3 | 439 | 59 | |
XM_012180607.3 | XP_012035997.1 | 3 - 4 | 805 | 59 | ||
XM_027971440.1 | XP_027827241.1 | 3 - 4 | 752 | 59 | ||
GenBank mRNA | AF508312.1 | - | 1 - 2 | 362 | 59 | |
DQ152931.1 | - | 1 - 2 | 70 | 59 | ||
EU183348.1 | - | 2 - 3 | 439 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |