Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TARBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004006291.4 | XP_004006340.1 | 1 - 2 | 428 | 97 | |
XM_012174155.3 | XP_012029545.1 | 1 - 2 | 434 | 97 |