Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OLFML2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004002705.4 | XP_004002754.1 | 6 - 7 | 2014 | 77 | |
XM_012185028.3 | XP_012040418.1 | 6 - 7 | 2021 | 77 | ||
XM_012185031.3 | XP_012040421.1 | 6 - 7 | 2018 | 77 | ||
XM_012185041.3 | XP_012040431.1 | 6 - 7 | 1972 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |