Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCGR3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001139454.1 | NP_001132926.1 | 4 - 5 | 592 | 77 | |
XM_012188046.2 | XP_012043436.2 | 4 - 5 | 627 | 77 | ||
XM_012188574.2 | XP_012043964.2 | 4 - 5 | 624 | 77 | ||
GenBank mRNA | EU589390.1 | - | 4 - 5 | 592 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Ovis aries |