Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYOZ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199222.1 | NP_001186151.1 | 2 - 3 | 153 | 119 | |
GenBank mRNA | HM628579.1 | - | 2 - 3 | 153 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |