Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001246210.1 | NP_001233139.1 | 1 - 2 | 213 | 99 | |
GenBank mRNA | FJ915124.1 | - | 1 - 2 | 213 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Ovis aries |