Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001131030.1 | NP_001124502.1 | 2 - 3 | 289 | 119 | |
GenBank mRNA | EU797604.1 | - | 2 - 3 | 289 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Ovis aries |