Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127275.1 | NP_001120747.1 | 4 - 5 | 455 | 73 | |
XM_027957545.1 | XP_027813346.1 | 4 - 5 | 770 | 73 | ||
GenBank mRNA | EU626218.1 | - | 4 - 5 | 455 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Ovis aries |