Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NANOG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004006901.4 | XP_004006950.3 | 1 - 2 | 136 | 84 | |
XM_015094959.2 | XP_014950445.1 | 1 - 2 | 136 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Ovis aries |