Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012106011.2 | XP_011961401.2 | 6 - 7 | 669 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Ovis aries |