Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MANEA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004011265.4 | XP_004011314.2 | 4 - 5 | 869 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |