Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RUNDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004012954.4 | XP_004013003.1 | 2 - 3 | 653 | 97 | |
XM_012186140.3 | XP_012041530.1 | 3 - 4 | 659 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Ovis aries |