Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RUNDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004012954.4 | XP_004013003.1 | 4 - 5 | 976 | 60 | |
XM_012186140.3 | XP_012041530.1 | 5 - 6 | 982 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Ovis aries |