Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LXN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004003214.4 | XP_004003263.2 | 5 - 6 | 844 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Ovis aries |