Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027960115.1 | XP_027815916.1 | 5 - 6 | 1457 | 93 | |
XM_027960116.1 | XP_027815917.1 | 4 - 5 | 533 | 93 | ||
XM_027960117.1 | XP_027815918.1 | 5 - 6 | 1460 | 93 | ||
XM_027960118.1 | XP_027815919.1 | 5 - 6 | 1459 | 93 | ||
XM_027960119.1 | XP_027815920.1 | 5 - 6 | 1461 | 93 | ||
XM_027960120.1 | XP_027815921.1 | 4 - 5 | 1363 | 93 |