Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD79A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004015292.4 | XP_004015341.2 | 3 - 4 | 664 | 71 | |
XM_012190460.3 | XP_012045850.1 | 3 - 4 | 472 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Ovis aries |